ESD safety shoes, MADDOX BOA®, S3
Toimittaja: ELTEN
729151/35 729151/36 729151/37 729151/38 729151/39 729151/40 729151/41 729151/42 729151/43 729151/44 729151/45 729151/46 729151/47 729151/48
114-8889EA 106.9 EUR
114-8889 114-8890 114-8891 114-8892 114-8893 114-8894 114-8895 114-8896 114-8897 114-8898 114-8899 114-8900 114-8901 114-8902
ESD safety shoes, MADDOX BOA®, S3
Kengät ja saappaat
Ideal for indoor or outdoor use, in areas with an exposure to moisture, a risk of penetration from pointed and sharp objects, or a risk of electrostatic discharge.
  • Hydrophobised microfibre/hydrophobised textile material
  • Fabric lining for natural ventilation
  • Padded tongue
  • Full length inlay sole SPORTIVE ESD black
  • Non metallic, penetration-resistant midsole
  • TPU/PU sole MAXXIMO
  • Steel toe cap
  • BOA® Fit System
  • Leather-free equipment, sole core made of Infinergy® by BASF
Equipped with the quick and practical BOA® Fit System, shoes come without laces, buckles,More Product Information