ESD safety ankle boots, lace-Up, L10, shadow mid
Toimittaja: ELTEN
761081/37 761081/39 761081/40 761081/41 761081/42 761081/44 761081/45 761081/46 761081/47
114-8800EA 93.9 EUR
114-8800 114-8802 114-8803 114-8804 114-8805 114-8807 114-8808 114-8809 114-8810
ESD safety ankle boots, lace-Up, L10, shadow mid
Kengät ja saappaat
Casual, vintage style, environmental friendly manufactured ESD safety ankle boot made from cowhide leather, a full padded bellows tongue, leather lining and a dual density TPU/PU sole (L10).
  • Very low weight helps to lightweight upper and sole materials
  • Anatomically shaped insole and ankle support for improved stability
  • Replaceable insole; orthopaedic insoles possible
  • Full-length inlay sole ESD PRO beige
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where thereMore Product Information