ESD safety shoes, lace-Up, Wellmax, Sensation XXT
Toimittaja: ELTEN
729720/36 729720/37 729720/46 729720/38 729720/39 729720/41 729720/42 729720/44 729720/43 729720/45 729720/40 729720/48 729720/47
114-8760EA 91.9 EUR
114-8760 114-8761 114-8770 114-8762 114-8763 114-8765 114-8766 114-8768 114-8767 114-8769 114-8764 114-8772 114-8771
ESD safety shoes, lace-Up, Wellmax, Sensation XXT
Kengät ja saappaat
Environmental friendly manufactured ESD safety shoes made from hydrophobised textile material in 'denim look' and hydrophobised nubuck leather, a padded tongue plus a low profile, dual-density PU/PU sole WELLMAXX TRAINERS.
  • Bioactive fabric lining
  • Full-length inlay sole ESD PRO grey
  • Anatomically shaped insole and ankle support for improved stability
  • Cushioning midsole made of Infinergy® by BASF rebounds back extremely well when under pressure giving the energy back to the wearer
  • Metal-free, puncture resistant midsole
  • Composite toecap (200 J)