ESD safety shoes, lace-up, Senex XXT
Toimittaja: ELTEN
729731/36 729731/37 729731/38 729731/39 729731/40 729731/41 729731/42 729731/43 729731/44 729731/45 729731/46 729731/47 729731/48 729731/35 729731/49
114-8475EA 96.9 EUR
114-8475 114-8476 114-8477 114-8478 114-8479 114-8480 114-8481 114-8482 114-8483 114-8484 114-8485 114-8486 114-8487 ELTN729731/35 ELTN729731/49
ESD safety shoes, lace-up, Senex XXT
Kengät ja saappaat
Metal- and leather-free, sporty, lightweight ESD safety shoe made from hydrophobised microfibre and hydrophobised CORDURA® (tear and abrasion resistant, dirt and water repellent), a padded tongue plus a slip resistant, dual-density TPU/PU sole Trainers.
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole ESD Pro black
  • TPU scuff cap protects the leather upper
  • Metal-free, puncture resistant midsole
  • Composite toecap (200 J)
  • Very wide version
Ideal for use in areas where exposure toMore Product Information