ESD safety shoes, lace-up, Miss L10, Vintage Lady Low
Toimittaja: ELTEN
74101/35 74101/36 74101/37 74101/38 74101/39 74101/40 74101/41 74101/42
114-7731EA 85.9 EUR
114-7731 114-7732 114-7733 114-7734 114-7735 114-7736 114-7737 114-7738
ESD safety shoes, lace-up, Miss L10, Vintage Lady Low
Kengät ja saappaat
Casual, vintage style, environment friendly manufactured ESD safety shoe made from waxy cowhide leather, a padded tongue plus a low profile, dual-density TPU/PU sole (Miss L10).
  • Bioactive fabric lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole (ESD Pro), grey
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects. The shoe is tailored to the ergonomics of female feet.