ESD safety ankle boots, Velcro® fastening, uvex 1, 8517
Toimittaja: UVEX
8517.2/35 8517.2/36 8517.2/37 8517.2/38 8517.2/39 8517.2/40 8517.2/41 8517.2/42 8517.2/43 8517.2/44 8517.2/45 8517.2/46 8517.2/47 8517.2/48 8517.2/49 8517.2/50 8517.2/51 8517.2/52 8517.1/52 8517.1/38 8517.1/51 8517.1/36 8517.1/41 8517.1/44 8517.1/47 8517.1/35 8517.1/40 8517.1/37 8517.1/42 8517.1/45 8517.1/39 8517.1/46 8517.1/48 8517.1/50 8517.1/49 8517.1/43
114-7189EA 148 EUR
114-7189 114-7190 114-7191 114-7192 114-7193 114-7125 114-7126 114-7127 114-7128 114-7129 114-7130 114-7131 114-7132 114-7133 114-7134 114-7135 114-7136 114-7137 UVEX8517.1/52 UVEX8517.1/38 UVEX8517.1/51 UVEX8517.1/36 UVEX8517.1/41 UVEX8517.1/44 UVEX8517.1/47 UVEX8517.1/35 UVEX8517.1/40 UVEX8517.1/37 UVEX8517.1/42 UVEX8517.1/45 UVEX8517.1/39 UVEX8517.1/46 UVEX8517.1/48 UVEX8517.1/50 UVEX8517.1/49 UVEX8517.1/43
ESD safety ankle boots, Velcro® fastening, uvex 1, 8517
Kengät ja saappaat
Metal-free, extremely lightweight, flexible and multifunctional safety shoe made from breathable water resistant high-tech micro velour upper, soft padded tongue and collar, plus a dual-density PUR sole (oil and petrol resistant, anti-static) free of lacquer-wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Anti-slip sole with shock-absorption at heel and front of the foot
  • Virtually