ESD-työkengät, pistokkaat, Dynamic, 37331 to 37335
Toimittaja: Abeba
37331/41 37335/42 37335/41 37335/40 37335/39 37335/38 37335/37 37335/36 37335/35 37335/43 37335/47 37335/45 37335/44 37335/48 37335/46 37331/42 37332/35 37331/48 37331/47 37331/46 37331/45 37332/37 37331/43 37332/38 37331/44 37332/45 37332/48 37332/36 37332/46 37332/44 37332/43 37332/42 37332/41 37332/40 37332/39 37332/47 37331/40 37331/39 37331/38 37331/37 37331/36 37331/35
114-6138EA 66.4 EUR
114-6138 114-8025 114-8024 114-8023 114-8022 114-8021 114-8020 114-8019 114-8018 114-8026 114-8030 114-8028 114-8027 114-8031 114-8029 114-6139 114-6160 114-6145 114-6144 114-6143 114-6142 114-6162 114-6140 114-6163 114-6141 114-6170 114-6173 114-6161 114-6171 114-6169 114-6168 114-6167 114-6166 114-6165 114-6164 114-6172 114-6137 114-6136 114-6135 114-6134 114-6133 114-6132
ESD-työkengät, pistokkaat, Dynamic, 37331 to 37335
Kengät ja saappaat
ESD clog made from smooth microfibre with a non foldable, adjustable heel strap and a TPU sole (resistant to fuel, oil and animal fats).
  • Replaceable acc Wave insole; orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • TPU sole with high slip resistance
  • Washable at 30 °C
Colour: Black/blue, black/red, black/yellow, white/black
Sertifikaatit: EN 61340 (ESD); HACCP; DGUV 112-191; EN ISO 20347:2012; OB, FO,E,A, SRC.