Turvanilkkurit, nauhoilla, Mac Crossroad
Toimittaja: UVEX
6322346 6322347 6322348 6322337 6322339 6322344 6322336 6322340
114-5928EA 0 EUR
114-5928 114-5929 114-5930 114-5919 114-5921 114-5926 114-5918 114-5922
Turvanilkkurit, nauhoilla, Mac Crossroad
Kengät ja saappaat
100% metal-free multipurpose safety ankle boot made from liquid resistant fullgrain leather and a MACSOLE® rubber sole (resistant to oils, hydrocarbons and chemicals, heat resistant up to 300 °C).
  • Mesh lining
  • Removable, anatomically shaped CELL TECH hygienic insole: Perforated and shock absorbing
  • Two component rubber sole with excellent abrasion and cut resistance and self-cleaning profile
  • Metal-free, lightweight toecap (200 J)
  • Metal-free puncture resistant midsole
  • Rubber scuffcap, reinforced heel and sides
For uneven terrain and humid environments.