Turvanilkkurit, nauhoilla, Athletic Ascender Mid
Toimittaja: HONEYWELL SAFETY
6243851/37 6243851/38 6243851/39 6243851/40 6243851/41 6243851/42 6243851/43 6243851/44 6243851/45 6243851/46 6243851/47
114-5509EA 176 EUR
114-5509 114-5510 114-5511 114-5512 114-5513 114-5514 114-5515 114-5516 114-5517 114-5518 114-5519
Turvanilkkurit, nauhoilla, Athletic Ascender Mid
Kengät ja saappaat
Safety ankle boots made from water resistant leather and textile upper, 3-D lining, watertight bellows plus PU/TPU sole.
  • Breathable, absorbent, fast drying Poromax® 3-D lining
  • Coolmax® insole for an optimal perspiration absorption
  • Shock-absorbent PU/TPU sole
  • Metal-free, flexible, puncture resistant FLEXIUM ™ midsole with thermal insulation
  • Ultra-light 200 J Athletic™ metal-free toecap
  • Forefoot and heel reinforcements
  • Speed lacing system and secured lace pocket
Colour: Black/grey
Colour: Black/grey
Sertifikaatit: EN ISO 20345:2004; S3, CI, SRC