Alumiini ≥99.998% (metals basis), folio, Puratronic®, Thickness: 1.0 mm (0.04 in)

Toimittaja: Thermo Fisher Scientific

11373.FI 11373.GH 11373.HP
11373.FIEA 175 EUR
11373.FI 11373.GH 11373.HP
Alumiini ≥99.998% (metals basis), folio, Puratronic®, Thickness: 1.0 mm (0.04 in)
Alumiini

Purity: 99.998% CAS No.: 7429-90-5 Physical Form: ≈6.75g/50x50mm, Temper: as rolled Odour: Odourless MDL No.: MFCD00134029

Kaava: Al
Kiehumispiste: 2327 °C (1013 hPa)
Sulamispiste: 660 °C
Tiheys: 2,7 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00134029
CAS-numero: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR