Titaanidiboridi

Toimittaja: Thermo Fisher Scientific

11364.18 11364.30 11364.A1
11364.18EA 131 EUR
11364.18 11364.30 11364.A1
Titaanidiboridi
Titaanidiboridi

CAS No.: 12045-63-5 Molecular Formula: B2Ti Formula Weight: 69.50 Physical Form: -325 Mesh Powder Density: 4.38 Odour: Odourless MDL No.: MFCD00049595


In the aircraft industry, in cutting and grinding applications, as protective coatings for metals in high temperature applications, as a base material for cermet production

Kaava: B₂Ti
Sulamispiste: 3225 °C
Tiheys: 4,38
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00049595
CAS-numero: 12045-63-5
EINECS: 234-961-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR