Takki, puhdastila, Quantus® Protect Xtra
Toimittaja: VWR Collection
113-9942EA 0 EUR
113-9942 113-9943 113-9944 113-9945 113-9946 113-9947 113-9948 113-9949 113-9950
Takki, puhdastila, Quantus® Protect Xtra
Työvaatteet
These coats are made from Protect Xtra fabric which is a further development of the Protect fabric consisting of 100% polyester weave with conductive yarn (warp). This fabric has been especially designed for use in critical areas (e.g. operating rooms, sterile environments). Thanks to the PTFE finish, the coats provide very good splash protection against liquids and other contamination. The fabric has good tensile strength and provides very good resistance to microbial penetration.
  • Lapped seams ensure well-sealed and secure seams
  • Press-stud closure, high
More Product Information
Order Now
TEKNISET TIEDOT
Environmentally Preferable
Asu sovellukset Cleanroom,Medical,Clinical
Risk area Anti-static
Sukupuoli Unisex
Tulpan tyyppi Painonapeilla
Kangas 100% polyester weave with conductive yarn
Paino 98,3 g/m²