VWR®, Launderable cleanroom socks
Toimittaja: VWR Collection
113-9939EA 1630 EUR
113-9939 113-9936 113-9937 113-9938
VWR®, Launderable cleanroom socks
Työvaatteet
Cleanroom socks provide comfort and durability for limited and extended wear in cleanroom environments.
  • 100% polyester
  • Colour coded bands for visual identification
  • Cleanroom compatible
  • No particle generation
  • Launderable
Minimise the risk of particle contamination with VWR® cleanroom socks.
Ideal for use with VWR® cleanroom clogs.
These launderable cleanroom socks provide comfort for extended wear and are cleanroom compatible.
Independent tests recommend that VWR launderable socks can withstand a minimum of 60 laundering cycles with less than 1More Product Information