Visitor coats, PP, velcro
Toimittaja: VWR Collection
113-9276EA 0 EUR
113-9276 113-9283 113-9272 113-9273 113-9277 113-9278 113-9280 113-9282
Visitor coats, PP, velcro
Kertakäyttöiset vaatteet
Disposable visitor coats made from PP.
  • Front closure with four velcro and collar
  • No pocket
  • Latex-free elastic wrist
Weight: 35 g/m²
Pakkaus: One per bag and 50 per carton.