VWR® Basic, Aprons, PE
Toimittaja: VWR Collection
113-9102EA 77.9 EUR
113-9102 113-9100 113-9101 113-9096 113-9095 113-9099 113-9097 113-9098 113-9104 113-9106 113-9103
VWR® Basic, Aprons, PE
Kertakäyttöiset vaatteet
Disposable aprons made of PE film.
  • Free from natural rubber latex
  • Available loose or blocked
L×W: 1250×800 mm / 1400×800 mm / 1600×800 mm
Thickness: 20, 50 or 100 µm