VWR® Basic, Frocks, Zip Closure
Toimittaja: VWR Collection
113-9000EA 94.2 EUR
113-9000 113-9001
VWR® Basic, Frocks, Zip Closure
Kertakäyttöiset vaatteet
These disposable frocks are manufactured from a strong, lightweight and breathable, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime and certain dry particulates in non hazardous environments.
  • Zip closure
  • Elastic wrists
  • Raglan sleeves
  • Free from natural rubber latex
Weight: 40 g/m²
Pakkaus: 1 piece per bag, 50 bags per case.