Chemical resistant overalls, high visibility, Tyvek® 500 HV
Toimittaja: DuPont
TY125SHVSM0025 TY125SHVMD0025 TY125SHVLG0025 TY125SHVXL0025 TY125SHV2X0025 TY125SHV3X0025
113-8720EA 602 EUR
113-8720 113-8719 113-8718 113-8723 113-8721 113-8722
Chemical resistant overalls, high visibility, Tyvek® 500 HV
Kertakäyttöiset vaatteet
These overalls are mainly composed of flash spun polyethylene, providing an ideal balance of protection, durability and comfort. Thanks to their non woven structure, they are permeable to both air and water vapour, yet repel water-based liquids and aerosols. The overalls also offer an excellent barrier against fine particles and fibres.
  • Ultra-low linting fabric
  • Anti-static treatment (EN 1149-5) on inside
  • Stitched external seams
  • Very low inward leakage thanks to optimised design
  • With collar, Tyvek® auto-lock zip and zip flap for increased protection