Bouffant caps, Tyvek® IsoClean®, model IC 729 S
Toimittaja: DuPont
IC729SWH000 IC729SWHMS IC729SWH00
DUPOIC729SWH000EA 0 EUR
DUPOIC729SWH000 113-8646 DUPOIC729SWH00
Bouffant caps, Tyvek® IsoClean®, model IC 729 S
Kertakäyttöiset vaatteet
These caps are made of Tyvek® which is a fabric composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash.
  • Ultra-low linting fabric
  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Elastic headband
  • Garments are