Cleanroom boot covers, Tyvek® IsoClean®, model IC458B MS
Toimittaja: DuPont
IC458BWHMS/L IC458BWHMS/XL IC458BWHMS/S IC458BWHMS/M
113-8642EA 353 EUR
113-8642 113-8643 113-8640 113-8641
Cleanroom boot covers, Tyvek® IsoClean®, model IC458B MS
Kertakäyttöiset vaatteet
These sterile overboots are made of Tyvek®. This fabric is composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort.
  • Ultra-low-linting fabric
  • Fixation ties
  • Ankle ties for secure fit
  • Gripper™ soles provide enhanced skid-resistance and durability
  • Clean laundered and gamma-sterilised
  • Sterility assurance level (SAL) of 10⁻⁶
  • Bound seams: tightly sewn seam is covered with garment fabric to reinforce seam protection and reduce potential for liquid and particle penetration
Tyvek® is permeable to bothMore Product Information