Huomiotakki, sade, valokaarisuojauksella, Nash 7227
Toimittaja: SIOEN
7227LK2143/S 7227LK2143/M 7227LK2143/L 7227LK2143/3XL 7227LK2143/2XL 7227LK2143/1XL
113-7247EA 447 EUR
113-7247 113-7246 113-7245 113-7244 113-7243 113-7242
Huomiotakki, sade, valokaarisuojauksella, Nash 7227
Työvaatteet
This high performance jacket is made of a 2-layer laminated fabric consisting of 60% FR Modacryl and 40% cotton (& AST) ripstop with PU laminate. The jacket features a fixed flame-retardant cotton lining but other fleeces and lining can be zipped in at any time.
  • Highly breathable, taped seams
  • Straight collar, provided for hood (hood available separately)
  • Zip closure under flap with touch and close fastening
  • One patched chest pocket, two patched pockets, two loops for badges, Napoleon pocket
  • Round inset sleeves, narrowing by touch and close fastening