Sadetakki, parka, 4-in-1, Cloverfield 288A
Toimittaja: SIOEN
288A/201/1XL 288A/201/2XL 288A/201/3XL 288A/201/L 288A/201/M 288A/201/S 288A/061/1XL 288A/061/2XL 288A/061/3XL 288A/061/L 288A/061/M 288A/061/S 288A/201/4XL 288A/061/4XL
113-7079EA 166 EUR
113-7079 113-7080 113-7081 113-7083 113-7084 113-7085 113-7066 113-7067 113-7068 113-7070 113-7071 113-7072 113-7082 113-7069
Sadetakki, parka, 4-in-1, Cloverfield 288A
Työvaatteet
Multifunctional breathable jacket made of Siopor® Ultra (100% polyester fabric with 100% PU coating) that can be used in all weather conditions. The outer fabric is water-repellent and windproof while the coating on the inside is moisture attracting. With fixed lining, knitted wind cuffs in sleeves and taped seams.
  • Detachable bodywarmer, detachable sleeves
  • Straight collar with foldaway hood
  • Zip closure under flap with touch & close fastening
  • Two inset pockets, Napoleon pocket, Mica pocket, inside pockets in lining and bodywarmer
  • Kimono sleeves with