Hihansuoja, kemikaalinkestävä, Tychem® C ja Tychem® F
Toimittaja: DuPont
TYCPS32SYL00 TYFPS32SGY00
113-0879EA 152 EUR
113-0879 113-0883
Hihansuoja, kemikaalinkestävä, Tychem® C ja Tychem® F
Kertakäyttöiset vaatteet
Specially designed for use with Tychem® apparel, these sleeve protectors can help offer enhanced protection for body parts that are more exposed to hazardous substances.
  • Wide elastics at cuffs and upper arm
  • Stitched and over-taped seams with barrier tape for protection and strength
  • Anti-static treatment (EN 1149-5) on inside
Tychem® 2000 C garments utilise the strength of Tyvek® and a polymeric barrier coating to offer good permeation barrier protection against a wide range of inorganic chemicals and biological hazards (even under pressure). IdeallyMore Product Information