Boot covers, Tyvek® 500, models POBA/POB0
Toimittaja: DuPont
TYPOB0SWH00020 TYPOBASWH00020
113-0489EA 416 EUR
113-0489 113-0490
Boot covers, Tyvek® 500, models POBA/POB0
Kertakäyttöiset vaatteet
Specially designed for use with Tyvek® apparel, these knee-length boot covers can help offer enhanced protection for body parts that are more exposed to hazardous substances.
  • Ultra-low linting fabric
  • Elasticated top and fixation ties
  • Stitched internal seams
  • Anti-static treatment (EN 1149-5) on both sides
  • With or without slip-retardant sole
Tyvek® garments are composed of flash spun high density PE, providing an ideal balance of protection, durability and comfort. The fabric is permeable to both air and water vapour, yet repels water-based liquids andMore Product Information