Suojatakki, puhdastila, Tyvek® 500
Toimittaja: DuPont
TYPL30SWHLG53P TYPL30SWHMD53P TYPL30SWHXL53P TYPL30SWH2X53P TYPL30SWHLG52P TYPL30SWHMD52P TYPL30SWHSM005 TYPL30SWHXL52P TYPL30SWH2X52P TYPL30SWHLG5NP TYPL30SWHMD5NP TYPL30SWHXL5NP TYPL30SWH2X5NP
113-0467EA 459 EUR
113-0467 113-0468 113-0469 113-0470 113-0471 113-0472 113-3882 113-0473 113-0474 113-0475 113-0476 113-0477 113-0478
Suojatakki, puhdastila, Tyvek® 500
Kertakäyttöiset vaatteet
These garments are composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 µm in size).
  • Fabric is ultra-low linting
  • With collar and stitched internal seams
  • Anti-static treatment (EN 1149-5) on both sides
  • PL30 with straight cuffs, PL309 + PL30NP with elasticated cuffs (not tunnelled)
  • Available with zip or press stud closure, with