Työhousut, pitkät, Fristads® PR54-220, Design B, musta
Toimittaja: FRISTADS KANSAS
84094013152 84094013148 84094013150 84094013154 84094013156 84094013158 84094013160 84094013162 840940210146 840940210148 840940210150 840940210152 840940210154 840940210156 84094021044 84094021046 84094021048 84094021050 84094021052 84094021054 84094021056 84094021058 84094021060 84094021062 840940231146 840940231148 840940231150 840940231152 840940231154 840940231156 84094023144 84094023146 84094023148 84094023150 84094023152 84094023154 84094023156 84094023158 84094023160 84094023162 840940331146 840940331148 840940331150 840940331152 840940331154 840940331156 84094033144 84094033146 84094033148 84094033150 84094033152 84094033154 84094033156 84094033158 84094033160 84094033162 840940440146 840940440148 840940440150 840940440152 840940440154 840940440156 84094044044 84094044046 84094044048 84094044050 84094044052 84094044054 84094044056 84094044058 84094044060 84094044062 840940520146 840940520148 840940520150 840940520152 840940520154 840940520156 84094052044 84094052046 84094052048 84094052050 84094052052 84094052054 84094052056 84094052058 84094052060 84094052062 840940530146 840940530148 840940530150 840940530152 840940530154 840940530156 84094053044 84094053046 84094053048 84094053050 84094053052 84094053054 84094053056 84094053058 84094053060 84094053062 840940539146 840940539148 840940539150 840940539152 840940539154 840940539156 84094053944 84094053946 84094053948 84094053950 84094053952 84094053954 84094053956 84094053958 84094053960 84094053962 84094064044 84094064046 84094064048 84094064050 84094064052 84094064054 840940131146 840940131148 840940131150 840940131152 840940131154 840940131156 84094013144 84094013146 84094064056 84094064058 84094064060 84094064062 840940730146 840940730148 840940730150 840940730152 840940730154 840940730156 84094073044 84094073046 84094073048 84094073050 84094073052 84094073054 84094073056 84094073058 84094073060 84094073062 840940940146 840940940148 840940940150 840940940152 840940940154 840940940156 84094094044 84094094046 84094094048 84094094050 84094094052 84094094054 84094094056 84094094058 84094094060 840940941146 840940941148 840940941150 840940941152 840940941154 840940941156 84094094144 84094094146 84094094148 84094094150 84094094152 84094094154 84094094156 84094094158 84094094160 84094094162
113-01499EA 0 EUR
113-01499 113-01497 113-01498 113-01500 113-01501 113-01502 113-01503 113-01504 113-03041 113-03042 113-03043 113-03044 113-03045 113-03046 113-01475 113-01476 113-01477 113-01478 113-01479 113-01480 113-01481 113-01482 113-01483 113-01484 113-03059 113-03060 113-03061 113-03062 113-03063 113-03064 113-01505 113-01506 113-01507 113-01508 113-01509 113-01510 113-01511 113-01512 113-01513 113-01514 113-03065 113-03066 113-03067 113-03068 113-03069 113-03070 113-01515 113-01516 113-01517 113-01518 113-01519 113-01520 113-01521 113-01522 113-01523 113-01524 113-03071 113-03072 113-03073 113-03074 113-03075 113-03076 113-01525 113-01526 113-01527 113-01528 113-01529 113-01530 113-01531 113-01532 113-01533 113-01534 113-03011 113-03012 113-03013 113-03014 113-03015 113-03016 113-01425 113-01426 113-01427 113-01428 113-01429 113-01430 113-01431 113-01432 113-01433 113-01434 113-03023 113-03024 113-03025 113-03026 113-03027 113-03028 113-01445 113-01446 113-01447 113-01448 113-01449 113-01450 113-01451 113-01452 113-01453 113-01454 113-03017 113-03018 113-03019 113-03020 113-03021 113-03022 113-01435 113-01436 113-01437 113-01438 113-01439 113-01440 113-01441 113-01442 113-01443 113-01444 113-01455 113-01456 113-01457 113-01458 113-01459 113-01460 113-03053 113-03054 113-03055 113-03056 113-03057 113-03058 113-01495 113-01496 113-01461 113-01462 113-01463 113-01464 113-03035 113-03036 113-03037 113-03038 113-03039 113-03040 113-01465 113-01466 113-01467 113-01468 113-01469 113-01470 113-01471 113-01472 113-01473 113-01474 113-03077 113-03078 113-03079 113-03080 113-03081 113-03082 113-01535 113-01536 113-01537 113-01538 113-01539 113-01540 113-01541 113-01542 113-01543 113-03047 113-03048 113-03049 113-03050 113-03051 113-03052 113-01485 113-01486 113-01487 113-01488 113-01489 113-01490 113-01491 113-01492 113-01493 113-01494
Työhousut, pitkät, Fristads® PR54-220, Design B, musta
Työvaatteet
Thanks to their durability and comfort, these trousers are perfectly suitable for use in garages as well as in industrial and service companies. The material is made of 65% polyester and 35% cotton. The inside is brushed and provides a comfortable cotton feel. The outside is very hardwearing and does not bleach, even after countless washes.
  • Two front and back pockets
  • Two leg pockets - one with flap, telephone pocket, pen pocket, tool bags
  • Adjustable waist, plastic snap buttons
  • Colour accents on pocket edges and leg pocket flap
Weight: 260 g/m²
Basic colour: black