Holmiumtrikloridihexahydraatii ≥99.9% (REO, rare earth oxide basis), REacton®

Toimittaja: Thermo Fisher Scientific
Varoitus

11277.14 11277.22
11277.14EA 340 EUR
11277.14 11277.22
Holmiumtrikloridihexahydraatii ≥99.9% (REO, rare earth oxide basis), REacton®
Holmiumtrikloridihexahydraatii

Purity: 99.9% CAS No.: 14914-84-2 Molecular Formula: Cl3Ho.6H2O Formula Weight: 379.38 (271.29anhy) Storage Temperature: 2°C to 8°C Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic Odour: Odourless Melting Point: 718° Merck Reference: 14,4727 MDL No.: MFCD00149739

Kaava: Cl₃Ho·6H₂O
Sulamispiste: 718 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00149739
CAS-numero: 14914-84-2
EINECS: 233-387-1
Merck Index: 14,4727

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR