Lanthanum(III) fluoride, vedetön ≥99.99% (REO, rare earth oxide basis), REacton®

Toimittaja: Thermo Fisher Scientific
Varoitus

11273.18 11273.30
11273.18EA 349 EUR
11273.18 11273.30
Lanthanum(III) fluoride, vedetön ≥99.99% (REO, rare earth oxide basis), REacton®
Lanthanum(III) fluoride

Purity: 99.99% CAS No.: 13709-38-1 Molecular Formula: F3La Formula Weight: 195.90 Physical Form: Crystalline Density: 5.936 Melting Point: 1493° MDL No.: MFCD00011073

Kaava: F₃La
Kiehumispiste: ∼2327 °C (1013 hPa)
Sulamispiste: 1493 °C
Tiheys: 5,936
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011073
CAS-numero: 13709-38-1
EINECS: 237-252-8
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR