Lanthanum, compound with nickel (1:5) ≥99.9% (REO, rare earth oxide basis), REacton®

Toimittaja: Thermo Fisher Scientific

11271.18 11271.90
11271.18EA 280 EUR
11271.18 11271.90
Lanthanum, compound with nickel (1:5) ≥99.9% (REO, rare earth oxide basis), REacton®
Lanthanum, compound with nickel (1:5)

Lanthanum nickel act as a superconducting intermetallic phase in lanthanum nickel boro-nitride.


Purity: 99.9% CAS No.: 12196-72-4 Molecular Formula: LaNi5 Physical Form: Pieces, Intermetallic, Packaged under argon Sensitivity: Air and Moisture Sensitive Odour: Odourless UN No.: 3178 MDL No.: MFCD00198177

Kaava: LaNi₅
MDL numero: MFCD00198177
CAS-numero: 12196-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR