Praseodyymitrifluoridi, vedetön ≥99.9895% (REO, rare earth oxide basis), REacton®

Toimittaja: Thermo Fisher Scientific
Vaara

11241.09 11241.18
11241.09EA 363 EUR
11241.09 11241.18
Praseodyymitrifluoridi, vedetön ≥99.9895% (REO, rare earth oxide basis), REacton®
Praseodyymitrifluoridi

Purity: 99.9895% CAS No.: 13709-46-1 Molecular Formula: F3Pr Formula Weight: 197.90 Physical Form: Powder Density: 6.2 Appearance: Green Odour: Odourless Melting Point: 1395° UN No.: 3288 MDL No.: MFCD00016287

Kaava: F₃Pr
Sulamispiste: 1395 °C
Tiheys: 6,2
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00016287
CAS-numero: 13709-46-1
EINECS: 237-254-9
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR