Praseodyymi(III,IV)oksidi ≥99.5% (REO, rare earth oxide basis)

Toimittaja: Thermo Fisher Scientific
Varoitus

11234.14 11234.22 11234.A1
11234.14EA 245 EUR
11234.14 11234.22 11234.A1
Praseodyymi(III,IV)oksidi ≥99.5% (REO, rare earth oxide basis)
Praseodyymi(III,IV)oksidi

Purity: 99.5% CAS No.: 12037-29-5 Molecular Formula: Pr6O11 Formula Weight: 1021.43 Physical Form: Powder Melting Point: 2500° MDL No.: MFCD00011178

Kaava: O₁₁Pr₆
Sulamispiste: 2500 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011178
CAS-numero: 12037-29-5
EINECS: 234-857-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR