Terbium(III) fluoride, vedetön ≥99.99% (REO, rare earth oxide basis), REacton®

Toimittaja: Thermo Fisher Scientific
Varoitus

11213.06 11213.14
11213.06EA 333 EUR
11213.06 11213.14
Terbium(III) fluoride, vedetön ≥99.99% (REO, rare earth oxide basis), REacton®
Terbium(III) fluoride

Purity: 99.99% CAS No.: 13708-63-9 Molecular Formula: TbF3 Formula Weight: 215.92 Physical Form: Crystalline Sensitivity: Hygroscopic UN No.: 3288 MDL No.: MFCD00049572

Kaava: TbF₃
Kiehumispiste: 2277 °C (1013 hPa)
Sulamispiste: 1172 °C
Tiheys: 7,23 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00049572
CAS-numero: 13708-63-9
EINECS: 237-247-0
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR