Terbium(III)kloridihexahydraatii ≥99.99% (REO, rare earth oxide basis), REacton®

Toimittaja: Thermo Fisher Scientific
Varoitus

11210.09 11210.18
11210.09EA 352 EUR
11210.09 11210.18
Terbium(III)kloridihexahydraatii ≥99.99% (REO, rare earth oxide basis), REacton®
Terbium(III)kloridihexahydraatii

Purity: 99.99% CAS No.: 13798-24-8 Molecular Formula: TbCl3.6H2O Formula Weight: 373.38 (265.29anhy) Physical Form: Crystalline Aggregates Odour: Odourless Merck Reference: 14,9157 MDL No.: MFCD00149870

Kaava: Cl₃Tb·6H₂O
Sulamispiste: 145 °C
Tiheys: 4,35 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149870
CAS-numero: 13798-24-8
EINECS: 233-132-4
Merck Index: 14,9157

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR