Terbium(III,IV)oksidi ≥99.9% (REO, rare earth oxide basis), REacton®

Toimittaja: Thermo Fisher Scientific

11207.09 11207.18 11207.A1 11207.30
11207.09EA 665 EUR
11207.09 11207.18 11207.A1 11207.30
Terbium(III,IV)oksidi ≥99.9% (REO, rare earth oxide basis), REacton®
Terbium(III,IV)oksidi

Purity: 99.9% CAS No.: 12037-01-3 Molecular Formula: Tb4O7 Formula Weight: 747.69 Physical Form: Powder Odour: Odourless Merck Reference: 14,9157 MDL No.: MFCD00083151

Kaava: O₇Tb₄
Sulamispiste: 2340 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00083151
CAS-numero: 12037-01-3
EINECS: 234-856-3
Merck Index: 14,9157

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR