Nitrile gloves, Showa 7502PF
Toimittaja: Showa
7502PFL 7502PFM 7502PFS 7502PFXL 7502PFXS 7502PFXXL
112-6594EA 53.3 EUR
112-6594 112-6595 112-6596 112-6597 112-6598 112-6599
Nitrile gloves, Showa 7502PF
Käsineet
Designed for sensitive skin, the 7502PF nitrile disposable glove is powder-, latex- and accelerator-free.
  • Biodegradable
  • Ambidextrous
  • 0,06 mm nitrile provides good chemical protection from a wide array of chemical hazards
  • Large dispenser size: 180/200 gloves per dispenser
Featuring SHOWA’s revolutionary Eco Best Technology (EBT), these gloves are engineered for accelerated biodegradation in biologically active landfills.
Recommended for: Handling plants and vegetables, light chemical handling in horticulture, agriculture, food packing and handling,More Product Information