Kimtech™ Polaris™ Nitrile Gloves
Toimittaja: KIMBERLY CLARK
62000 62001 62002 62003 62004
112-6560EA 29.3 EUR
112-6560 112-6561 112-6562 112-6563 112-6564
Kimtech™ Polaris™ Nitrile Gloves
Käsineet
Kimtech™ Polaris™ Nitrile Gloves are built for science, made and designed for use in laboratories, life science applications, and non-sterile pharmaceutical manufacturing environments.
  • High-end ambidextrous powder-free nitrile gloves deliver seamless protection whenever and wherever it is needed
  • Optimized nitrile formulation (acrylonitrile-butadiene) features extended chemicals and chemotherapy drugs splash protection without compromising comfort
  • Beaded cuffs and textured fingertips enable excellent handling of both wet and dry materials
  • Extensively tested