Nitrile gloves, white, PUREZERO® HG3
Toimittaja: Halyard
CLN323265 CLN323270 CLN323275 CLN323280 CLN323285 CLN323290 CLN3031LG CLN3031MD CLN3031SM CLN3031XL CLN3031XS CLN323210 CLN323260
112-6526EA 528 EUR
112-6526 112-6527 112-6528 112-6529 112-6530 112-6531 112-6509 112-6510 112-6511 112-6512 112-6513 112-6524 112-6525
Nitrile gloves, white, PUREZERO® HG3
Käsineet
PUREZERO® nitrile cleanroom gloves are made by an accelerator-free formulation which reduces the risk of allergies and skin irritation associated with accelerator chemicals in other nitrile gloves. As a result, PUREZERO® gloves are comfortable to wear, allowing workers to focus on their delicate tasks rather than their gloves.
  • Designed to protect workers with effective barrier protection against chemical splash, microorganisms and viruses
  • Clean-processed to ensure consistent control of low particles and extractables
  • Recommended for use in ISO Class 3
More Product Information
Order Now
TEKNISET TIEDOT
Tyyppi HG3
Environmentally Preferable
Molempiin käsiin sopiva/käsispesifinen Molempiin käsiin sopiva
Lateksi/Lateksiton Lateksiton
Puuteroitu/Puuteroimaton Talkiton
Pinta Textured fingers
Rannekkeen paksuus 4 mil
Sormen paksuus 6 mil
Kämmenen paksuus 5 mil
Murtovenymä 600%
EN Normi EN ISO 374-1:2016/Type C (K), EN ISO 374-5:2016, EN ISO 374-4