Cleanroom gloves, nitrile, non sterile, white, smooth finish, PUREZERO® HG3
Toimittaja: Halyard
CLN3231LG CLN3231XL CLN3231MD CLN3231SM CLN3231XS
112-6519EA 331 EUR
112-6519 112-6522 112-6520 112-6521 112-6523
Cleanroom gloves, nitrile, non sterile, white, smooth finish, PUREZERO® HG3
Käsineet
High quality gloves are designed to exact standards to meet the requirements of a contamination controlled environment.
  • High quality nitrile
  • Non sterile
  • Accelerator-free formula reduces risk of type IV allergies
  • For use in cleanroom environments (ISO 5/6/7/8, grade B/C/D aseptic)
  • REACH Annex XVII
Offering the right glove for the right controlled environment, Halyard PUREZERO® provides a cleanroom glove with guaranteed low particulate counts and endotoxin levels due to repeated washes in deionised water and chlorination. Nickel-, powder-, silicon- andMore Product Information