Cleanroom Latex Gloves, Non Sterile, LS300
Toimittaja: ISOFIELD IRELAND
10230XS 10230S 10230M 10230L 10230XL 10230XXL
112-6482EA 354 EUR
112-6482 112-6480 112-6479 112-6478 112-6481 112-6483
Cleanroom Latex Gloves, Non Sterile, LS300
Käsineet
The Isofield LS300 non-sterile latex glove is ideal for ISO Class 4 cleanrooms and above.
  • ISO Class 4, GMP Grade A compatible
  • Non sterile cleanroom latex
  • 300 mm length
  • Low endotoxin
  • Ambidextrous
Triple washed in de-ionized water, the LS300 is smooth enough for double donning while also offering a textured palm and fingers for added grip.
Sertifikaatit: PPE Cat III, Type B certified.
Pakkaus: 100 pieces, double packaged in HDPE bag, 10 bags per case (1000 pieces).