Cleanroom Nitrile Gloves, Extended Cuff, Non Sterile, NS400
Toimittaja: ISOFIELD IRELAND
20340S 20340M 20340L 20340XL
112-6451EA 638 EUR
112-6451 112-6450 112-6469 112-6452
Cleanroom Nitrile Gloves, Extended Cuff, Non Sterile, NS400
Käsineet
The Isofield NS400 non-sterile nitrile extended cuff glove offers excellent elbow length protection for ISO Class 4 cleanrooms and above.
  • ISO Class 4 compatible
  • Non sterile, nitrile gloves
  • Extra length (400 mm)
  • Low endotoxin
  • Anti-static
  • Tested against Chemotherapy
Triple washed in de-ionized water, the NS400 is smooth enough for double donning while also offering a textured palm and fingers for added grip.
Sertifikaatit: PPE Cat III, Type B certified.