Cleanroom Nitrile Hand Specific Gloves, Sterile, Volta
Toimittaja: ISOFIELD IRELAND
2053060 2053065 2053070 2053075 2053080 2053085 2053090 20530100
112-6462EA 577 EUR
112-6462 112-6463 112-6464 112-6465 112-6466 112-6467 112-6468 112-6461
Cleanroom Nitrile Hand Specific Gloves, Sterile, Volta
Käsineet
The Isofield Volta sterile green nitrile glove offers excellent protection for GMP Grade A, ISO Class 4 cleanrooms and above.
  • ISO Class 4 compatible
  • Sterile cleanroom nitrile
  • 300 mm length
  • Low endotoxin
  • Hand specific
  • Double-donnable
  • Anti-static
Perfect as an under glove, the triple DI washed, low endotoxin Volta glove is anti-static, and double-donnable.
Sertifikaatit: PPE Cat III, Type B certified.
Pakkaus: Each pair is double packaged in a wallet within a pouch, 10 pouches per bag, 20 bags per case (200 pairs).