Cleanroom gloves, latex, sterile, Kimtech™ G3
Toimittaja: KIMBERLY CLARK
56843 56844 56845 56846 56847 56848 56849 56842
112-6413EA 418 EUR
112-6413 112-6412 112-6411 112-6410 112-6409 112-6408 112-6407 112-6414
Cleanroom gloves, latex, sterile, Kimtech™ G3
Käsineet
These sterile latex gloves are designed to ensure high levels of contamination control in critical and controlled cleanroom environments.
  • High levels of contamination control in critical and controlled cleanroom environments
  • Sterilised latex ensures compliant and seamless protection when and where it counts
  • High-quality finish, with enhanced texturing on the palm and fingertips, provides improved grip, tactile sensitivity and comfort in both wet and dry conditions
  • Hand-specific gloves also feature a beaded cuff that provides added strength and makes