VWR® VINYL HYBRID, Gloves, Vinyl
Toimittaja: VWR Collection
112-6072EA 16.9 EUR
112-6072 112-6073
VWR® VINYL HYBRID, Gloves, Vinyl
Käsineet
A glove lower in price than nitrile and higher in quality than vinyl.
  • Non sterile
  • Powder-free
  • Particularly skin friendly for users with latex allergy
  • Rolled cuffs minimise the risk of tearing
These innovative vinyl gloves are made from a revolutionary new formula of vinyl-based hybrid material. The formulation combines vinyl and nitrile properties in one glove by using the best of each material and providing outstanding value. These gloves offer enhanced tactile sensitivity and good barrier protection for a wide range of applications, such as foodMore Product Information