Nitrile gloves, Showa 6110PF
Toimittaja: Showa
6110PFS 6110PFL 6110PFXL 6110PFXXL 6110PFM 6110PFXS
112-5637EA 31.2 EUR
112-5637 112-5639 112-5640 112-5641 112-5638 BEST6110PFXS
Nitrile gloves, Showa 6110PF
Käsineet
These disposable gloves are powder-free and made of 100% nitrile. By using Showa’s Eco Best Technology®, the 6110PF’s are completely eco-friendly, breaking down by 82% in just 386 days when tested in a laboratory.
  • Zero natural rubber latex proteins
  • Rolled cuff
  • Decreased risk of allergies
  • High performance of protection
  • 'Second skin' fit and feel
Eco Best Technology® (EBT) is a breakthrough innovation that makes the gloves completely, without affecting shelf life or sacrificing performance. When disposed in landfills, microorganisms consume, metaboliseMore Product Information