VWR® Cleanroom Gloves, Soft Nitrile, Non Sterile, ISO 4
Toimittaja: VWR Collection
112-5145EA 427 EUR
112-5145 112-5146 112-5147 112-5148 112-5150 112-5149
VWR® Cleanroom Gloves, Soft Nitrile, Non Sterile, ISO 4
Käsineet
VWR® cleanroom soft, nitrile gloves are manufactured from carboxylated acrylonitrile butadiene that provides reliable, durable and comfortable hand protection during demanding cleanroom applications.
  • Ambidextrous, textured finger and beaded cuff
  • Not made from natural rubber latex
  • Static-dissipative
  • Single-use only
  • New and unique formulation to prevent or minimise Type IV allergies, or dermatitis potential, caused by the use of sulphur and most accelerators in conventional formulations
  • High elongation, and softer feel for extra comfort during donning,