Käsineet, aluskäsineet, HyFlex® 11-318
Toimittaja: Ansell
11-318/11 11-318/10 11-318/9 11-318/6 11-318/7 11-318/8
112-4718EA 193 EUR
112-4718 112-4717 112-4722 112-4719 112-4720 112-4721
Käsineet, aluskäsineet, HyFlex® 11-318
Käsineet
This uncoated glove provides EN Level 3 cut protection in an 18 gauge liner. Its ultra light fabric, made of Dyneema® Diamond, Spandex, nylon and carbon fibres, creates a sensation of bare hand contact and helps to maintain the hand at a comfortable working temperature.
  • Snug/ergonomic fit on the hand thanks to the spandex reinforcement in the tension areas of the hand (knuckles and palm)
  • Low linting, silicone- and latex-free and anti-static
  • Specific packaging in order to prevent silicone contamination
Ideal for the final assembly of sharp parts, forMore Product Information