Käsineet, kemikaalinkestävät, PowerCoat® 080-10 Butyl
Toimittaja: HONEYWELL SAFETY
2008010/10 2008010/8 2008010/9 2008010/11
112-1584EA 0 EUR
112-1584 112-1588 112-1589 DALL2008010/11
Käsineet, kemikaalinkestävät, PowerCoat® 080-10 Butyl
Käsineet
These unsupported butyl gloves offer resistance to many solvents, acids and medium concentrated bases as well as to ozone and heat. They are also gas tight.
  • Remains supple from −30 up to +50 °C
  • Butyl is more flexible than natural latex, tear resistant and provides good dexterity
  • Smooth inside to ease donning and duffing
  • Rolled edge stops drops and reinforces the gauntlet resistance to tear
Ideal for offshore and petro-chemical applications as well as for water treatments.
Thickness: 0,5 mm
Colour: Black
EN 388 (3010), EN 374 (BKL), Cat. III