Chemical resistant gloves, AlphaTec® Solvex 37-675
Toimittaja: Ansell
37-675/7 37-675/8 37-675/9 37-675/10 37-675/11 37-675/6
112-1380EA 62.7 EUR
112-1380 112-1381 112-1382 112-0210 112-0966 112-0967
Chemical resistant gloves, AlphaTec® Solvex 37-675
Käsineet
These nitrile gloves with cotton flock lining are designed to deliver optimal results in wet or dry work environments where chemical resistance is crucial.
  • Fully reusable, very good abrasion resistance
  • Cotton flock lining combined with the flexibility of the nitrile film offer exceptional comfort for the wearer
  • Sandpatch finish gives the glove a smooth surface that decreases indirect costs thanks to fewer rejects of fragile parts
  • Antistatic according to EN 1149
Typical applications include: Agrochemicals, chemical processing and preparation, metalMore Product Information
Order Now
TEKNISET TIEDOT
Tyyppi 37-675
Molempiin käsiin sopiva/käsispesifinen Käsispesifinen
Environmentally Preferable
Kertakäyttöinen/uudelleenkäytettävä Uudelleenkäytettävä
Vuoren materiaali Puuvilla
Ranneke Pitkä, suora
EN Normi EN ISO 374-1:2016 Type A (JKLOPT), EN ISO 374-5:2016 VIRUS, EN 388:2016 (4101X)
Antisaattisuus +
Hyväksytty elintarvikekäyttöön +