Käsineet, puhdastila, nitriili, BioClean Excell™
Toimittaja: Nitritex
BEXS-100 BEXS-60 BEXS-65 BEXS-70 BEXS-75 BEXS-80 BEXS-85 BEXS-90
112-1320EA 665 EUR
112-1320 112-1324 112-1334 112-1339 112-1340 112-1341 112-1347 112-1348
Käsineet, puhdastila, nitriili, BioClean Excell™
Käsineet
These nitrile gloves with beaded cuffs are hand-specific and anatomically shaped.
  • Sterile, powder-free, textured surface
  • Contain no natural latex proteins, hypoallergenic
  • Extended leaching in pure de-ionised water ensures extremely low levels of chlorides and extractable metals
  • Nitrile is resistant to a wide range of chemicals
  • Excellent anti-static properties
Suitable for use in Class 10, ISO 4 or M2.5 cleanrooms.
Typical particle count ≥0,5 μm: <1200 counts/cm²
Sertifikaatit: Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425More Product Information