Käsineet, kemikaalinkestävät, AlphaTec® Butyl, 38-514 / 38-520
Toimittaja: Ansell
38-514/10 38-514/7 38-514/8 38-514/9 38-520/10 38-520/9 38-520/8 38-520/7 38-520/11
112-3781EA 85.7 EUR
112-3781 112-3778 112-3779 112-3780 112-1269 112-1268 112-1570 112-1569 ANSE38-520/11
Käsineet, kemikaalinkestävät, AlphaTec® Butyl, 38-514 / 38-520
Käsineet
Maximum protection from dangerous chemicals. Made from Butyl these gloves offer effective protection against a considerably wider range of aggressive industrial chemicals than conventional glove materials, such as nitrile, neoprene and latex.
  • Freedom of movement, comfort and excellent grip
  • Designed for comfort – with a natural, curved ergonomic shape and soft feel
  • Latex-free formulation: No risk of latex-related allergic reactions
  • Effective protection against aldehydes, ketones and esters, as well as concentrated mineral acids
Length: 350 mm