Käsineet, puhdastila, nitriili, Nitrilite® 93-311
Toimittaja: Ansell
93-311/6.5-7 93-311/7.5-8 93-311/8.5-9 93-311/9.5-10
112-0455EA 388 EUR
112-0455 112-0456 112-1165 112-0461
Käsineet, puhdastila, nitriili, Nitrilite® 93-311
Käsineet
These ambidextrous silicone-free gloves are produced in a controlled environment. They feature exceptionally low levels of extractables and particles for excellent product protection.
  • Non sterile, powder-free, textured fingertips
  • Free of latex proteins, preventing the risk of allergies
  • Excellent puncture and chemical resistance
  • Optimal electrical properties
Suitable for use in Class 100, ISO 5 or M3.5 cleanrooms.
Ideal for a wide range of industrial sectors, from electronics to paint shops in automotive manufacturing and biotechnologies.